5-Iod-o-toluidin
Lieferant: CHEMPUR
Synonyme:
2-Amino-4-iodtoluol, 5-Iod-2-methylanilin
MB00009.250
CHMPMB00009.250EA
0
EUR
CHMPMB00009.250
5-Iod-o-toluidin
5-Iod-o-toluidin
Formel:
C₇H₈IN Molecular Weight: 233,05 g/mol Siedepunkt: 273 °C (1013 hPa) Schmelzpunkt: 48…49 °C |
MDL:
MFCD00051531 CAS-Nummer: 83863-33-6 EINECS: 281-094-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...