3,5-Dimethoxybenzonitril
Lieferant: CHEMPUR
Synonyme:
3,5-Dimethoxybenzoesäurenitril
MB00259.250
CHMPMB00259.250EA
0
EUR
CHMPMB00259.250
3,5-Dimethoxybenzonitril
3,5-Dimethoxybenzonitril
Formel:
C₉H₉NO₂ Molecular Weight: 163,18 g/mol Siedepunkt: 283 °C (1013 hPa) Schmelzpunkt: 92 °C |
MDL:
MFCD00001804 CAS-Nummer: 19179-31-8 EINECS: 242-858-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...