2,3-Dichlorchinoxalin
Lieferant: CHEMPUR
MGH00028.250
CHMPMGH00028.250EA
0
EUR
CHMPMGH00028.250
2,3-Dichlorchinoxalin
2,3-Dichloroquinoxaline
Formel:
C₈H₄Cl₂N₂ Molecular Weight: 199,04 g/mol Siedepunkt: 301 °C (1013 hPa) Schmelzpunkt: 152…153 °C |
MDL:
MFCD00006720 CAS-Nummer: 2213-63-0 EINECS: 218-667-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...