Carbohydrazid

Lieferant: CHEMPUR

ML00094.250
CHMPML00094.250EA 0 EUR
CHMPML00094.250
Carbohydrazid
Carbohydrazid
Formel: CH₆N₄O
Molecular Weight: 90,09 g/mol
MDL: MFCD00007591
CAS-Nummer: 497-18-7
EINECS: 207-837-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR