2-(1H-Pyrazol-1-yl)anilin

Lieferant: CHEMPUR

Synonyme: 2-(1-Pyrazolyl)aniline, 1-(2-Aminophenyl)-1H-pyrazole, 1-(2-Aminophenyl)pyrazole,2-(1H-Pyrazol-1-yl)aniline

MO00205.10
CHMPMO00205.10EA 0 EUR
CHMPMO00205.10
2-(1H-Pyrazol-1-yl)anilin
2-(1H-Pyrazol-1-yl)anilin
Formel: C₉H₉N₃
Molecular Weight: 159,1879 g/mol
MDL: MFCD03086227
CAS-Nummer: 54705-91-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR