4-Amino-2,6-dichlorpyridin
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
2,6-Dichlor-4-pyridylamin, 2,6-Dichlorpyridin-4-amin
RF04500.250
RF04500DA
CHMPRF04500.250EA
0
EUR
CHMPRF04500.250
MAYBRF04500DA
4-Amino-2,6-dichlorpyridin
4-Amino-2,6-dichlorpyridin
Formel:
C₅H₄Cl₂N₂ Molecular Weight: 163,01 g/mol |
MDL:
MFCD00052832 CAS-Nummer: 2587-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...