2-(Trifluormethoxy)anilin Hydrochlorid

Lieferant: CHEMPUR

RF05450.250 RF05450.1
CHMPRF05450.250EA 0 EUR
CHMPRF05450.250 CHMPRF05450.1
2-(Trifluormethoxy)anilin Hydrochlorid
2-(Trifluormethoxy)anilin Hydrochlorid
Formel: C₇H₇ClF₃NO
MDL: MFCD00219957
CAS-Nummer: 452077-64-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR