3,5-Dichlor-4-hydroxybenzaldehyd

Lieferant: CHEMPUR

RH00194.250 RH00194DA
CHMPRH00194.250EA 0 EUR
CHMPRH00194.250 MAYBRH00194DA
3,5-Dichlor-4-hydroxybenzaldehyd
3,5-Dichloro-4-hydroxybenzaldehyde
Formel: C₇H₄Cl₂O₂
MDL: MFCD00017605
CAS-Nummer: 2314-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR