6-Bromveratraldehyd

Lieferant: CHEMPUR

Synonyme: 6-Bromo-3,4-dimethoxybenzaldehyde, 2-Bromo-4,5-dimethoxybenzaldehyde

RH00868.250 RH00868.1
CHMPRH00868.250EA 0 EUR
CHMPRH00868.250 MAYBRH00868.1
6-Bromveratraldehyd
6-Bromveratraldehyd
Formel: C₉H₉BrO₃
Molecular Weight: 245.07 g/mol
Siedepunkt: 300
Schmelzpunkt: 151
MDL: MFCD00003301
CAS-Nummer: 5392-10-9
EINECS: 226-390-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR