5-Fluor-2-indolcarbonsäure

Lieferant: CHEMPUR

Synonyme: 5-Fluoro-1H-indole-2-carboxylic acid

RH01182.250
CHMPRH01182.250EA 0 EUR
CHMPRH01182.250
5-Fluor-2-indolcarbonsäure
5-Fluoroindole-2-carboxylic acid
Formel: C₉H₆FNO₂
Molecular Weight: 179,15 g/mol
Schmelzpunkt: 258…259 °C
MDL: MFCD00005612
CAS-Nummer: 399-76-8
EINECS: 206-919-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR