4-Nitrobenzamid
Lieferant: CHEMPUR
Synonyme:
4-Nitrobenzoesäureamid
RH01465.250
RH01465.1
CHMPRH01465.250EA
0
EUR
CHMPRH01465.250
MAYBRH01465.1
4-Nitrobenzamid
4-Nitrobenzamid
Formel:
C₇H₆N₂O₃ Molecular Weight: 166.14 g/mol Schmelzpunkt: 198…202 °C Dichte: 1.59 g/cm³ (20 °C) |
MDL:
MFCD00007994 CAS-Nummer: 619-80-7 EINECS: 210-613-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...