2-Methyl-3,5-bis(trifluormethyl)anilin

Lieferant: CHEMPUR

RJC02733.0.1
CHMPRJC02733.0.1EA 0 EUR
CHMPRJC02733.0.1
2-Methyl-3,5-bis(trifluormethyl)anilin
2-Methyl-3,5-bis(trifluormethyl)anilin
Formel: C₉H₇F₆N
CAS-Nummer: 243128-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR