1,1,1-Trifluor-5,5-dimethyl-2,4-hexandion

Lieferant: Thermo Scientific

Synonyme: 5,5-Dimethyl-1,1,1-trifluorohexane-2,4-dione

RJC04016.1 RJC04016.250
MAYBRJC04016.1EA 0 EUR
MAYBRJC04016.1 CHMPRJC04016.250
1,1,1-Trifluor-5,5-dimethyl-2,4-hexandion
1,1,1-Trifluor-5,5-dimethyl-2,4-hexandion
Formel: C₈H₁₁F₃O₂
Molecular Weight: 196.17 g/mol
Siedepunkt: 178
Dichte: 1.129 g/cm³ (20 °C)
MDL: MFCD00040841
CAS-Nummer: 22767-90-4
EINECS: 245-211-0
UN: 1224
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR