3,4-Xylylisothiocyanat

Lieferant: CHEMPUR

Synonyme: 3,4-Dimethylphenyl isothiocyanate

S01746.250
CHMPS01746.250EA 0 EUR
CHMPS01746.250
3,4-Xylylisothiocyanat
3,4-Xylylisothiocyanat
Formel: C₉H₉NS
Molecular Weight: 163.24
MDL: MFCD00031536
CAS-Nummer: 19241-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR