Anthranilohydrazid

Lieferant: CHEMPUR

Synonyme: 2-Aminobenzhydrazid, 2-Aminobenzoesäurehydrazid, 2-Aminobenzol-1-carbohydrazid, 2-Aminobenzohydrazid

S01773.250 S01773.1
CHMPS01773.250EA 0 EUR
CHMPS01773.250 MAYBS01773.1
Anthranilohydrazid
Anthranilohydrazid
Formel: C₇H₉N₃O
Molecular Weight: 151.17 g/mol
Siedepunkt: 378
Schmelzpunkt: 123
MDL: MFCD00017056
CAS-Nummer: 1904-58-1
EINECS: 217-604-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR