3-(1,3-Oxazol-5-yl)anilin

Lieferant: CHEMPUR

Synonyme: 3-(5-Oxazolyl)aniline

SEW00990DA SEW00990.10
MAYBSEW00990DAEA 0 EUR
MAYBSEW00990DA CHMPSEW00990.10
3-(1,3-Oxazol-5-yl)anilin
3-(1,3-Oxazol-5-yl)anilin
Formel: C₉H₈N₂O
Molecular Weight: 160,1700 g/mol
MDL: MFCD00052191
CAS-Nummer: 157837-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR