2-(Trifluormethoxy)benzylamin

Lieferant: CHEMPUR
Gefahr

Synonyme: (2-(Trifluoromethoxy)phenyl)methanamine

SEW02173.250
CHMPSEW02173.250EA 0 EUR
CHMPSEW02173.250
2-(Trifluormethoxy)benzylamin
2-(Trifluormethoxy)benzylamin
Formel: C₈H₈F₃NO
Molecular Weight: 191,15 g/mol
Siedepunkt: 77 °C (12 mmHg)
MDL: MFCD00085176
CAS-Nummer: 175205-64-8
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR