2,6-Dichlor-4-(trifluormethyl)benzolsulfonylchlorid

Lieferant: CHEMPUR

SEW03506.250 SEW03506DA
CHMPSEW03506.250EA 0 EUR
CHMPSEW03506.250 MAYBSEW03506DA
2,6-Dichlor-4-(trifluormethyl)benzolsulfonylchlorid
2,6-Dichlor-4-(trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₂Cl₃F₃O₂S
MDL: MFCD00220929
CAS-Nummer: 175205-76-2
UN: 3261
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR