3,5-Dibrombenzamid

Lieferant: CHEMPUR

Synonyme: 3,5-Dibrombenzoesäureamid

SEW04141.1 SEW04141.10
CHMPSEW04141.1EA 0 EUR
CHMPSEW04141.1 CHMPSEW04141.10
3,5-Dibrombenzamid
3,5-Dibrombenzamid
Formel: C₇H₅Br₂NO
Molecular Weight: 278.93 g/mol
MDL: MFCD00178771
CAS-Nummer: 175205-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR