Chlor(diphenyl)silan

Lieferant: CHEMPUR

SILS03825.
CHMPSILS03825.EA 0 EUR
CHMPSILS03825.
Chlor(diphenyl)silan
Chloro(diphenyl)silane
Formel: C₁₂H₁₁ClSi
Molecular Weight: 218,76 g/mol
MDL: MFCD00045144
CAS-Nummer: 1631-83-0
EINECS: 216-634-8
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR