Dichlordiethylsilan

Lieferant: CHEMPUR

SILS05952.
CHMPSILS05952.EA 0 EUR
CHMPSILS05952.
Dichlordiethylsilan
Dichlorodiethylsilane
Formel: C₄H₁₀Cl₂Si
MDL: MFCD00000493
CAS-Nummer: 1719-53-5
EINECS: 217-005-0
UN: 1767
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR