Diethoxymethylsilan

Lieferant: CHEMPUR

Synonyme: Diethoxy(methyl)silane

SILS06260.
CHMPSILS06260.EA 0 EUR
CHMPSILS06260.
Diethoxymethylsilan
Diethoxymethylsilan
Formel: C₅H₁₄O₂Si
Molecular Weight: 134,25 g/mol
MDL: MFCD00026749
CAS-Nummer: 2031-62-1
EINECS: 217-982-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR