Dimethoxy(diphenyl)silan

Lieferant: CHEMPUR

Synonyme: Diphenyldimethoxysilan

SILS06828.
CHMPSILS06828.EA 0 EUR
CHMPSILS06828.
Dimethoxy(diphenyl)silan
Dimethoxy(diphenyl)silan
Formel: C₁₄H₁₆O₂Si
Molecular Weight: 244,37 g/mol
MDL: MFCD00025690
CAS-Nummer: 6843-66-9
EINECS: 229-929-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR