Dimethoxymethylvinylsilan

Lieferant: CHEMPUR

Synonyme: Dimethoxy(methyl)(vinyl)silane

SILS06829.
CHMPSILS06829.EA 0 EUR
CHMPSILS06829.
Dimethoxymethylvinylsilan
Dimethoxymethylvinylsilan
Formel: C₅H₁₂O₂Si
Molecular Weight: 132,23 g/mol
MDL: MFCD00129170
CAS-Nummer: 16753-62-1
EINECS: 240-816-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR