Dimethylphenylsilan

Lieferant: CHEMPUR

SILS07310.
CHMPSILS07310.EA 0 EUR
CHMPSILS07310.
Dimethylphenylsilan
Dimethylphenylsilan
Formel: C₈H₁₂Si
MDL: MFCD00008257
CAS-Nummer: 766-77-8
EINECS: 212-170-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR