Chlor(methyl)diphenylsilan

Lieferant: CHEMPUR

SILS11274.
CHMPSILS11274.EA 0 EUR
CHMPSILS11274.
Chlor(methyl)diphenylsilan
Chlor(methyl)diphenylsilan
Formel: C₁₃H₁₃ClSi
MDL: MFCD00000498
CAS-Nummer: 144-79-6
EINECS: 205-639-0
UN: 2987
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR