Methyl(trimethylsilyl)acetat

Lieferant: CHEMPUR

Synonyme: (Trimethylsilyl)essigsäuremethylester

SILS11950.
CHMPSILS11950.EA 0 EUR
CHMPSILS11950.
Methyl(trimethylsilyl)acetat
Methyl(trimethylsilyl)acetat
Formel: C₆H₁₄O₂Si
CAS-Nummer: 2916-76-9
EINECS: 220-845-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR