Phenyltriethoxysilan

Lieferant: CHEMPUR

Synonyme: Triethoxy(phenyl)silan

SILS12934.
CHMPSILS12934.EA 0 EUR
CHMPSILS12934.
Phenyltriethoxysilan
Phenyltriethoxysilan
Formel: C₁₂H₂₀O₃Si
Molecular Weight: 240,37 g/mol
Siedepunkt: 237 °C (1013 hPa)
Schmelzpunkt: 41 °C
Dichte: 0,996 g/cm³ (20 °C)
MDL: MFCD00009065
CAS-Nummer: 780-69-8
EINECS: 212-305-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR