Propyltrimethoxysilan

Lieferant: CHEMPUR

Synonyme: Trimethoxy(propyl)silan

SILS18062.
CHMPSILS18062.EA 0 EUR
CHMPSILS18062.
Propyltrimethoxysilan
Propyltrimethoxysilan
Formel: C₆H₁₆O₃Si
Molecular Weight: 164,28 g/mol
Siedepunkt: 142…143 °C (1013 hPa)
Dichte: 0,935 g/cm³ (20 °C)
MDL: MFCD00043026
CAS-Nummer: 1067-25-0
EINECS: 213-926-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR