p-Tolyltrimethylsilan

Lieferant: CHEMPUR

Synonyme: Trimethyl-p-tolylsilane, Trimethyl(p-tolyl)silane

SILS20450.
CHMPSILS20450.EA 0 EUR
CHMPSILS20450.
p-Tolyltrimethylsilan
p-Tolyltrimethylsilan
Formel: C₁₀H₁₆Si
Molecular Weight: 164,32 g/mol
MDL: MFCD00014850
CAS-Nummer: 3728-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR