Vinyltrimethylsilan

Lieferant: CHEMPUR

Synonyme: Trimethyl(vinyl)silane

SILS20480.
CHMPSILS20480.EA 0 EUR
CHMPSILS20480.
Vinyltrimethylsilan
Vinyltrimethylsilan
Formel: C₅H₁₂Si
Molecular Weight: 100,24 g/mol
Siedepunkt: 55 °C (1013 hPa)
Dichte: 0,7038 g/cm³ (20 °C)
MDL: MFCD00008606
CAS-Nummer: 754-05-2
EINECS: 212-042-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR