Tris(trimethylsilyl)borat

Lieferant: CHEMPUR

Synonyme: Tris(trimethylsiloxy)borate

SILS21025.
CHMPSILS21025.EA 0 EUR
CHMPSILS21025.
Tris(trimethylsilyl)borat
Tris(trimethylsilyl)borat
Formel: C₉H₂₇BO₃Si₃
Molecular Weight: 278,376 g/mol
MDL: MFCD00051588
CAS-Nummer: 4325-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR