Vinylferrocen

Lieferant: CHEMPUR

SILS21752.
CHMPSILS21752.EA 0 EUR
CHMPSILS21752.
Vinylferrocen
Vinylferrocen
Formel: C₁₂H₁₂Fe
MDL: MFCD00001434
CAS-Nummer: 1271-51-8
EINECS: 215-041-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR