2-Amino-4,6-dimethylnicotinamid

Lieferant: CHEMPUR

SP00089.1 SP00089.10
CHMPSP00089.1EA 0 EUR
CHMPSP00089.1 CHMPSP00089.10
2-Amino-4,6-dimethylnicotinamid
2-Amino-4,6-dimethylnicotinamid
Formel: C₈H₁₁N₃O
MDL: MFCD00138572
CAS-Nummer: 7144-20-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR