3-(Chlormethyl)-5-methylisoxazol 95%+

Lieferant: CHEMPUR

SPB01262.250 SPB01262DA
CHMPSPB01262.250EA 0 EUR
CHMPSPB01262.250 MAYBSPB01262DA
3-(Chlormethyl)-5-methylisoxazol 95%+
3-(Chloromethyl)-5-methylisoxazole
Formel: C₅H₆ClNO
Molecular Weight: 131,560 g/mol
MDL: MFCD00221166
CAS-Nummer: 35166-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR