3-(Trifluormethyl)pyridin

Lieferant: CHEMPUR

SPB05925.1 SPB05925.10
CHMPSPB05925.1EA 0 EUR
CHMPSPB05925.1 CHMPSPB05925.10
3-(Trifluormethyl)pyridin
3-(Trifluoromethyl)pyridine
Formel: C₆H₄F₃N
Molecular Weight: 147.1 g/mol
Siedepunkt: 110…113 °C (1013 hPa)
MDL: MFCD00100032
CAS-Nummer: 3796-23-4
EINECS: 223-268-2
UN: 1992
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR