4-Aminophenylboronsäurepinakolester

Lieferant: CHEMPUR

T01-BM11.25
CHMPT01-BM11.25EA 0 EUR
CHMPT01-BM11.25
4-Aminophenylboronsäurepinakolester
4-Aminophenylboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
MDL: MFCD02093721
CAS-Nummer: 214360-73-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR