Methyl-3-cyan-4-methoxybenzoat
Lieferant: Thermo Scientific
Synonyme:
Methyl 3-cyano-4-methoxybenzoate
TL00306DA
TL00306EA
TL00306.50
MAYBTL00306DAEA
0
EUR
MAYBTL00306DA
MAYBTL00306EA
CHMPTL00306.50
Methyl-3-cyan-4-methoxybenzoat
Methyl-3-cyan-4-methoxybenzoat
Formel:
C₁₀H₉NO₃ Molecular Weight: 191.19 g/mol |
MDL:
MFCD00052930 CAS-Nummer: 25978-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...