4-(Trifluormethoxy)benzoylchlorid
Lieferant: CHEMPUR
TL00396.250
CHMPTL00396.250EA
0
EUR
CHMPTL00396.250
4-(Trifluormethoxy)benzoylchlorid
4-(Trifluormethoxy)benzoylchlorid
Formel:
C₈H₄ClF₃O₂ Siedepunkt: 90…92 °C (15 mmHg) Dichte: 1,42 g/cm³ (20 °C) |
MDL:
MFCD00052329 CAS-Nummer: 36823-88-8 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...