2-Chlor-6-nitrobenzonitril

Lieferant: CHEMPUR

Synonyme: 6-Chloro-2-nitrobenzonitrile

TL00403.250 TL00403.1
CHMPTL00403.250EA 0 EUR
CHMPTL00403.250 MAYBTL00403.1
2-Chlor-6-nitrobenzonitril
2-Chlor-6-nitrobenzonitril
Formel: C₇H₃ClN₂O₂
MDL: MFCD00007203
CAS-Nummer: 6575-07-1
EINECS: 229-496-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR