2-Methyl-1-nitronaphthalin
Lieferant: CHEMPUR
XBX00241.250
XBX00241.1
CHMPXBX00241.250EA
0
EUR
CHMPXBX00241.250
MAYBXBX00241.1
2-Methyl-1-nitronaphthalin
2-Methyl-1-nitronaphthalene
Formel:
C₁₁H₉NO₂ Siedepunkt: 318 °C (1013 hPa) Schmelzpunkt: 81 °C |
MDL:
MFCD00003914 CAS-Nummer: 881-03-8 EINECS: 212-917-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...