Ammoniaklösung

Lieferant: CUSTOM MADE CHEMICALS LAB
Gefahr

Synonyme: Ammoniumhydroxid, Ammoniak

1004705.1000
CMPL1004705.1000EA 0 EUR
CMPL1004705.1000
Ammoniaklösung
Ammoniaklösung

The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for purified water.


Instruction for use: The solution bottle should be open for the minimum time required to dispense the solution. After use, the bottle should be tightly recapped and stored under normal laboratory conditions.


Shelf Life on Ship Date: 12 Months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.

Formel: NH₃
Molecular Weight: 35.05 g/mol
MDL: MFCD00066650
CAS-Nummer: 1336-21-6
UN: 2672
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR