Iod 0.05 mol/l (0.1 N) in wässriger Lösung

Lieferant: CUSTOM MADE CHEMICALS LAB

3002700
CMPL3002700EA 0 EUR
CMPL3002700
Iod 0.05 mol/l (0.1 N) in wässriger Lösung
Iod
Formel: I₂
Molecular Weight: 253,81 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011355
CAS-Nummer: 7553-56-2
EINECS: 231-442-4
UN: 3264
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR