Kaliumpermanganat 0.02 mol/l (0.1 N) aqueous solution Reag. Ph. Eur. 3005300
Lieferant: CUSTOM MADE CHEMICALS LAB
|
|
Synonyme:
Permanganic acid potassium salt
3005300
CMPL3005300EA
0
EUR
CMPL3005300
Kaliumpermanganat 0.02 mol/l (0.1 N) aqueous solution Reag. Ph. Eur. 3005300
Potassium permanganate
Formel:
KMnO₄ Molecular Weight: 158.03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011364 CAS-Nummer: 7722-64-7 EINECS: 231-760-3 UN: 3082 ADR: 9 III Merck Index: 12,07824 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...