Ammoniumnitrat 2%

Lieferant: CUSTOM MADE CHEMICALS LAB

Synonyme: Ammonsalpeter

909D.20K.W.L1
CMPL909D.20K.W.L1EA 0 EUR
CMPL909D.20K.W.L1
Ammoniumnitrat 2%
Ammoniumnitrat
Formel: NH₄NO₃
Molecular Weight: 80.04 g/mol
Siedepunkt: 230
Schmelzpunkt: 169
Lagertemperatur: Raumtemperatur
MDL: MFCD00011425
CAS-Nummer: 6484-52-2
EINECS: 229-347-8
Merck Index: 13,00538

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR