Methanol

Lieferant: CUSTOM MADE CHEMICALS LAB
Gefahr

Synonyme: Hydroxymethan, Methylalkohol, Holzalkohol

SB243.1000
CMPLSB243.1000EA 0 EUR
CMPLSB243.1000
Methanol
Methanol

Bestellinformation: Shelf life on ship date: 24 Months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.

Formel: CH₃OH
Molecular Weight: 32,04 g/mol
Siedepunkt: 64,7 °C
Schmelzpunkt: –98 °C
Dichte: 0,79 g/cm³
Flammpunkt: 11 °C
MDL: MFCD00004595
CAS-Nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR