Methylisothiocyanat, Sigma-Aldrich®

Lieferant: CUSTOM MADE CHEMICALS LAB
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

SB27280.100MG
CMPLSB27280.100MGEA 0 EUR
CMPLSB27280.100MG
Methylisothiocyanat, Sigma-Aldrich®
Methylisothiocyanat
Formel: C₂H₃NS
Molecular Weight: 73,12 g/mol
Siedepunkt: 117 °C (1013 hPa)
Schmelzpunkt: 31…37 °C
Dichte: 1,069 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00004818
CAS-Nummer: 556-61-6
EINECS: 209-132-5
UN: 2477
ADR: 6.1,I
Merck Index: 12,06165

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Yellow to Orange
Appearance (Form) Solid
Infrared spectrum Conforms to Structure
Purity (GC) ≥96.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR