Tyramin
Lieferant: CUSTOM MADE CHEMICALS LAB
Synonyme:
2-(4-Hydroxyphenyl)ethylamin, 4-Hydroxyphenethylamim, 4-(2-Aminoethyl)phenol
SB49364.100MG
CMPLSB49364.100MGEA
0
EUR
CMPLSB49364.100MG
Tyramin
Tyramin
Standards: ISO 17034, ISO 17025, ISO 9001.
Bestellinformation: Store in a refrigerator at temperatures between 2 to 8 °C.
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol |
MDL:
MFCD00008193 CAS-Nummer: 51-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...