Histamin Dihydrochlorid, white, powder
Lieferant: CUSTOM MADE CHEMICALS LAB
|
Gefahr
|
Synonyme:
Histamine 2HCl, 4-(2-Aminoethyl)-1H-imidazole dihydrochloride, 2-(1H-Imidazol-4-yl)ethan-1-amine dihydrochloride
SB49374.100MG
CMPLSB49374.100MGEA
0
EUR
CMPLSB49374.100MG
Histamin Dihydrochlorid, white, powder
Histamin Dihydrochlorid
Standards: ISO 17034, ISO 17025, ISO 9001.
Bestellinformation: Store in a refrigerator at temperatures between 2 to 8 °C.
Formel:
C₅H₁₁Cl₂N₃ Molecular Weight: 184.07 g/mol Schmelzpunkt: 247...249 °C Dichte: 0,63 g/cm³ |
MDL:
MFCD00012703 CAS-Nummer: 56-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...