2,4,6-Trichloranisol
Lieferant: CUSTOM MADE CHEMICALS LAB
|
Achtung
|
SB5500.100MG
CMPLSB5500.100MGEA
0
EUR
CMPLSB5500.100MG
2,4,6-Trichloranisol
2,4,6-Trichloranisol
Formel:
C₇H₅Cl₃O Siedepunkt: 241 °C |
MDL:
MFCD00000588 CAS-Nummer: 87-40-1 |
Spezifikation Testergebnisse
XLogP3 | 4 |
Hydrogen bond donor count | 0 |
Rotatable bond count | 0 |
Exact mass | 1 |
Monoisotopic mass | 209.940598 g/mol |
Topological polar surface area | 209.940598 g/mol |
Heavy atom count | 9.2 Ų |
Formal charge | 11 |
Complexity | 0 |
Isotope atom count | 119 |
Defined atom stereocenter count | 0 |
Undefined atom stereocenter count | 0 |
Defined bond stereocenter count | 0 |
Undefined bond stereocenter count | 0 |
Covalently-bonded unit count | 1 |
Compound is canonicalized | Yes |
Hydrogen bond acceptor count | 1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...